Learn More
Medchemexpress LLC HY-U00248A 1mg Medchemexpress, LMP744 hydrochloride CAS:308246-57-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYU00248A1MG
Encompass_Preferred
Medchemexpress, HY-U00248A 1mg LMP744 hydrochloride CAS:308246-57-3 LMP744 hydrochloride (MJ-III65 hydrochloride) is a DNA intercalator and Topoisomerase I (Top1) inhibitor with antitumor activity [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4434