Learn More
TARGETMOL CHEMICALS INC LMP744 hydrochloride 1MG
SDP
Catalog No. 502875919
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. LMP744 hydrochloride (NSC-706744 hydrochloride) is a DNA intercalator and Topoisomerase I (Top1) inhibitor with antitumor activity. Purity 98.44%
Catalog No. 50-287-5919 Supplier TARGETMOL CHEMICALS INC Supplier No. T11872L1MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. LMP744 hydrochloride (NSC-706744 hydrochloride) is a DNA intercalator and Topoisomerase I (Top1) inhibitor with antitumor activity. Purity 98.44%