Learn More
Medchemexpress LLC LMP744 hydrochloride | 308246-57-3 | 488.92 | 1 ML
SDP
Catalog No. 5000449514
Encompass_Preferred
LMP744 hydrochloride (MJ-III65 hydrochloride) is a DNA intercalator and Topoisomerase I (Top1) inhibitor with antitumor activity. It is intended for research use only.
  • Dose-dependent accumulation of cells in the S and G2 phases of the cell cycle
  • Antiproliferative activity against various human cell lines
  • Moderately active against human A253 and FaDu tumor xenografts in nude mice
Catalog No. 50-004-49514 Supplier Medchemexpress LLC Supplier No. HYU00248A10MM/1ML
LMP744 hydrochloride (MJ-III65 hydrochloride) is a DNA intercalator and Topoisomerase I (Top1) inhibitor with antitumor activity. It is intended for research use only.
  • Dose-dependent accumulation of cells in the S and G2 phases of the cell cycle
  • Antiproliferative activity against various human cell lines
  • Moderately active against human A253 and FaDu tumor xenografts in nude mice