Learn More
Medchemexpress LLC LMP744 hydrochloride | 308246-57-3 | 488.92 | 5 MG
SDP
Catalog No. 5000449588
Encompass_Preferred
LMP744 hydrochloride (MJ-III65 hydrochloride) is a DNA intercalator and Topoisomerase I (Top1) inhibitor exhibiting antitumor activity. It is a solid, light brown to brown in appearance, with a purity of 99.08%.
  • DNA intercalator
  • Topoisomerase I (Top1) inhibitor
  • Exhibits antitumor activity
  • Induces dose-dependent accumulation of cells in the S and G2 phases of the cell cycle
  • Moderately active against human A253 and FaDu tumor xenografts in vivo
Catalog No. 50-004-49588 Supplier Medchemexpress LLC Supplier No. HYU00248A5MG
LMP744 hydrochloride (MJ-III65 hydrochloride) is a DNA intercalator and Topoisomerase I (Top1) inhibitor exhibiting antitumor activity. It is a solid, light brown to brown in appearance, with a purity of 99.08%.
  • DNA intercalator
  • Topoisomerase I (Top1) inhibitor
  • Exhibits antitumor activity
  • Induces dose-dependent accumulation of cells in the S and G2 phases of the cell cycle
  • Moderately active against human A253 and FaDu tumor xenografts in vivo