Learn More
Eurofins Discoverx LMPTP-A, 250 µg. (14-619M)
Supplier:  Eurofins Discoverx 14619M
Encompass
The LMPTP-A product is purified N terminal 6xHis-tagged recombinant LMPTP-A enzyme. This enzyme is full length. It retains its binding and enzymatic activity and is intended for use in phosphatase assays. This enzyme is provided as 250 μg of 2.075 mg/mL frozen solution. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9066