Learn More
Aviva Systems Biology Corp LMX1A Antibody - middle region (ARP31999_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31999_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LMX1A. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human LMX1A. Alternate names: LMX-1, LMX1, LMX1.1, MGC87616ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54413