Learn More
Aviva Systems Biology Corp LMX1A Antibody - N-terminal region (ARP32673_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32673_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LMX1A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LMX1A. Alternate names: LMX-1, LMX1, LMX1.1, MGC87616ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54702