Learn More
Aviva Systems Biology Corp Lmx1a Antibody - middle region (ARP37428_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37428_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Lmx1a. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of mouse Lmx1a. Alternate names: Lmx1.1, MGC129356, MGC129357, dr, dreher, sstApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-630