Learn More
Aviva Systems Biology Corp LMX1B Antibody - C-terminal region (ARP38140_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38140_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against LMX1B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human LMX1B. Alternate names: NPS1, LMX1.2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53098