Learn More
Wells Lamont Industrial™ LN 13 A7 Cut Resistant Glove
Catalog No. 18064005 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-064-005 X-Large
18-064-002 Small
18-064-003 Medium
18-064-004 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-064-005 Supplier Wells Lamont Industrial™ Supplier No. LN13A7XL
Add to Cart
Add to Cart
LN 13 glove with ANSI Level A7 cut resistance. CFR 21 approved for food contact. Has antimicrobial fibers.
The Whizard™ Antimicrobial LN13 A7 Series offers hand protection at a high overall value. The combination of high-performance fibers and stainless steel filament create excellent tensile strength and superior ANSI Level A7 cut protection.The durable fibers provide strength, not bulk, and offer 13 gauge abrasion resistance.
  • The LN Series is available in various other gauges for light to heavy duty applications.
  • All gauges are ideal for handling of sharp surfaces, food preparation, and meat and poultry processing
Specifications
Construction Seamless Knit
Coating Material Uncoated
Color Salt and Pepper
Cut Resistance Rating ANSI A7
Hand Ambidextrous
Latex Free Yes
Material High Performance Fibers and Stainless Steel Filament
Product Type Cut-resistant Gloves
Size X-Large
Gender Unisex
Glove Cuff Style Elasticated
Lining Unlined
Puncture Rating ASTM F2878 LEVEL 0
Thumb Style Straight