Learn More
Wells Lamont Industrial™ LN 7 Cut Resistant Glove
Catalog No. 18063977 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-063-977 X-Large
18-063-974 Small
18-063-975 Medium
18-063-976 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-063-977 Supplier Wells Lamont Industrial™ Supplier No. 135652
Add to Cart
Add to Cart
Whizard™ LN7 cut resistant glove. Offers ANSI A6 cut resistance. 7 gauge with antimicrobial fibers. Made in the USA.
The Whizard™ Antimicrobial LN Series offers economically sound hand protection.
  • The combination of high performance fibers and stainless steel filament create excellent tensile strength and superior cut protection.
  • The durable fibers provide strength, not bulk, and offer high 7 gauge abrasion resistance.
  • The LN Series is available in various other gauges for light to heavy duty applications.
  • All gauges are ideal for handling of sharp surfaces, food preparation, and meat and poultry processing.
Specifications
Construction Seamless Knit
Coating Material Uncoated
Color Gray
Cut Resistance Rating ANSI A6
Hand Ambidextrous
Latex Free Yes
Material High Performance Fibers and Stainless Steel Filament
Product Type Cut-resistant Gloves
Size X-Large
Gender Unisex
Glove Cuff Style Elasticated
Lining Unlined
Puncture Rating ASTM F2878 LEVEL 0
Thumb Style Straight