Description
Use Thermo Scientific™ LN2 Measuring Stick with our LN2 cryopreservation equipment.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Description | LN2 Measuring Stick |
| Type | LN2 Measuring Device |
| For Use With (Equipment) | Locator, Locator Plus, CryoPlus |
Frequently Asked Questions (FAQs)
The LN2 measuring stick has a total length of ~114 cm (~45 inches) and the measuring portion of the stick is scaled to ~100 cm (40+ inch).