Learn More
ATCC LN-229; Glioblastoma; Human (Homo sapiens)
Catalog No. 502382001
Encompass
LN-229 is a cell line exhibiting epithelial-like morphology that was isolated in 1979 from the right frontal parieto-occipital cortex of a White, 60-year-old, female patient with glioblastoma. This cell line was deposited by N de Tribolet and can be used in neuroscience research.
Catalog No. 50-238-2001 Supplier ATCC Supplier No. CRL2611
LN-229 is a cell line exhibiting epithelial-like morphology that was isolated in 1979 from the right frontal parieto-occipital cortex of a White, 60-year-old, female patient with glioblastoma. This cell line was deposited by N de Tribolet and can be used in neuroscience research.