Learn More
Cayman Chemical LN6023hydrochlorIde 10mg
SDP
Catalog No. 502942424
Encompass_Preferred
A CXCR7 agonist; induces β-arrestin recruitment in a reporter assay using HEK293T cells (EC50 = 3.5 µM) but not in HEK293T cells expressing human CXCR4; reduces the surface levels of P-selectin in isolated and washed human platelets at 100 µM
Catalog No. 50-294-2424 Supplier Cayman Chemical Supplier No. 4028710 MG
A CXCR7 agonist; induces β-arrestin recruitment in a reporter assay using HEK293T cells (EC50 = 3.5 µM) but not in HEK293T cells expressing human CXCR4; reduces the surface levels of P-selectin in isolated and washed human platelets at 100 µM