Learn More
Medchemexpress LLC LNA-A(Bz) amidite | 206055-79-0 | MFCD31707505 | ≥95.0% | 885.94 | C48H52N7O8P | 100 MG
SDP
Catalog No. 5000284480
Encompass_Preferred
LNA-A(Bz) amidite is a locked nucleic acid (LNA) adenine phosphoramidite intended as a building block for solid-phase synthesis of modified oligonucleotides, including antisense oligonucleotides (ASOs). It is supplied for research use in DNA/RNA synthesis workflows and is provided with manufacturer documentation such as datasheets and batch certificates.
  • Locked nucleic acid adenine monomer for enhanced hybridization affinity.
  • Phosphoramidite chemistry compatible with standard automated oligonucleotide synthesis.
  • Suitable for synthesis of antisense oligonucleotides and other modified oligos.
  • Provided with molecular and batch data to support research applications.
  • Available in multiple pack sizes to accommodate small-scale and larger-scale synthesis.
Catalog No. 50-002-84480 Supplier Medchemexpress LLC Supplier No. HYW570887100MG
LNA-A(Bz) amidite is a locked nucleic acid (LNA) adenine phosphoramidite intended as a building block for solid-phase synthesis of modified oligonucleotides, including antisense oligonucleotides (ASOs). It is supplied for research use in DNA/RNA synthesis workflows and is provided with manufacturer documentation such as datasheets and batch certificates.
  • Locked nucleic acid adenine monomer for enhanced hybridization affinity.
  • Phosphoramidite chemistry compatible with standard automated oligonucleotide synthesis.
  • Suitable for synthesis of antisense oligonucleotides and other modified oligos.
  • Provided with molecular and batch data to support research applications.
  • Available in multiple pack sizes to accommodate small-scale and larger-scale synthesis.