Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC LNA-A(Bz) amidite | 206055-79-0 | MFCD31707505 | 95.0% | 885.94 g/mol | C48H52N7O8P | 50 MG
SDP

Catalog No. 5000284988
Encompass_Preferred

LNA-A(Bz) amidite is a locked nucleic acid phosphoramidite monomer used for the solid-phase synthesis of antisense and modified oligonucleotides. It provides a benzoyl-protected adenine nucleoside in a 4'-C-locked ribose and is supplied as a solid or as a DMSO solution for immediate use.

  • Can be used in antisense oligonucleotide (ASO) synthesis.
  • Purity 95.0%.
  • Molecular weight 885.94 g/mol; formula C48H52N7O8P.
  • Soluble in DMSO; 100 mg/mL; ultrasonic agitation may be required.
  • Appearance: white to off-white solid.
  • Storage: 4°C protected from light; stock solutions -80°C (6 months) or -20°C (1 month).
  • Available in solid sizes (50 mg, 100 mg, 250 mg) and 10 mM DMSO solutions.

Catalog No. 50-002-84988 Supplier Medchemexpress LLC Supplier No. HYW57088750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LNA-A(Bz) amidite is a locked nucleic acid phosphoramidite monomer used for the solid-phase synthesis of antisense and modified oligonucleotides. It provides a benzoyl-protected adenine nucleoside in a 4'-C-locked ribose and is supplied as a solid or as a DMSO solution for immediate use.

  • Can be used in antisense oligonucleotide (ASO) synthesis.
  • Purity 95.0%.
  • Molecular weight 885.94 g/mol; formula C48H52N7O8P.
  • Soluble in DMSO; 100 mg/mL; ultrasonic agitation may be required.
  • Appearance: white to off-white solid.
  • Storage: 4°C protected from light; stock solutions -80°C (6 months) or -20°C (1 month).
  • Available in solid sizes (50 mg, 100 mg, 250 mg) and 10 mM DMSO solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.