Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical LNA MeC(Bz) amidite | 100 g | CAS 206055-82-5 | MDL MFCD30475591

Catalog No. 502356691
Encompass

LNA MeC(Bz) amidite is a Amidite reagent (Subcategory: LNA Series) sold by WuXi TIDES. Offered in 100 g. Store at -20 °C. SDS available for reference.

Specifications
- CAS: 206055-82-5
- MDL: MFCD30475591
- InChIKey: XHILZPWJJJDDDY-HXAWEXODSA-N
- Molecular Weight: 875.959761998
- Molecular Formula: C48H54N5O9P
- Purity: ≥99%
- Container Type: 500 mL HDPE
- Pack Size: 100 g
- Net Weight: 100 g
- Gross Weight: 160.5 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: (1R,3R,4R,7S)-3-(4-benzamido-5-methyl-2-oxopyrimidin-1(2H)-yl)-1-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-2,5-dioxabicyclo[2.2.1]heptan-7-yl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: CC(C(NC(C1=CC=CC=C1)=O)=NC2=O)=CN2[C@@H]3O[C@]4(CO[C@@H]3[C@@H]4OP(N(C(C)C)C(C)C)OCCC#N)COC(C5=CC=C(C=C5)OC)(C6=CC=C(C=C6)OC)C7=CC=CC=C7

Catalog No. 50-235-6691 Supplier STA Pharmaceutical Supplier No. STAAM10093100GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LNA MeC(Bz) amidite is a Amidite reagent (Subcategory: LNA Series) sold by WuXi TIDES. Offered in 100 g. Store at -20 °C. SDS available for reference.

Specifications
- CAS: 206055-82-5
- MDL: MFCD30475591
- InChIKey: XHILZPWJJJDDDY-HXAWEXODSA-N
- Molecular Weight: 875.959761998
- Molecular Formula: C48H54N5O9P
- Purity: ≥99%
- Container Type: 500 mL HDPE
- Pack Size: 100 g
- Net Weight: 100 g
- Gross Weight: 160.5 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: (1R,3R,4R,7S)-3-(4-benzamido-5-methyl-2-oxopyrimidin-1(2H)-yl)-1-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-2,5-dioxabicyclo[2.2.1]heptan-7-yl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: CC(C(NC(C1=CC=CC=C1)=O)=NC2=O)=CN2[C@@H]3O[C@]4(CO[C@@H]3[C@@H]4OP(N(C(C)C)C(C)C)OCCC#N)COC(C5=CC=C(C=C5)OC)(C6=CC=C(C=C6)OC)C7=CC=CC=C7

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.