Learn More
TA Instruments LNCS and LN Pump DSC Purge Gas Purifier Assembly, Removes Trace Moisture in Purge Gases, Includes Shut-Off Valve, by-Pass Valve, Dryer Cartridge, Fittings, Provides Dry Gas to -180°C Dew Point
Catalog No. 5000580673 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments LNCS and LN Pump DSC Purge Gas Purifier Assembly is designed for removal of trace moisture in purge gases. The assembly includes a stainless steel shut-off valve, by-pass valve, gas dryer cartridge, and appropriate fittings. It provides dry gas to -180°C dew point.
Catalog No. 50-005-80673 Supplier TA Instruments Supplier No. 970425.901
Waters TA Instruments LNCS and LN Pump DSC Purge Gas Purifier Assembly is designed for removal of trace moisture in purge gases. The assembly includes a stainless steel shut-off valve, by-pass valve, gas dryer cartridge, and appropriate fittings. It provides dry gas to -180°C dew point.