Learn More
Med Vet International Safety Lancet McKesson 23 Gauge Retractable Push Button Activation Finger, 16-PBSL23GX, 100/Box, 20 Boxes
Catalog No. 503044304
Encompass_Preferred
Blue color-coded design for easy gauge identification Features gamma-sterilized 23-gauge needles, fully covered and protected with a sterility tab Reliability provided by optimized needle exposure of 1.8 mm fixed depth to guarantee adequate blood flow Electro-polished, silicone-coated needle with high puncture speed minimizes pain and facilitates healing for gentle finger tip sampling Easy-to-handle, retractable design features an intuitive, 2-step operation with low firing force and no lancet loading required
Catalog No. 50-304-4304 Supplier Med Vet International Supplier No. 1217990CS
Blue color-coded design for easy gauge identification Features gamma-sterilized 23-gauge needles, fully covered and protected with a sterility tab Reliability provided by optimized needle exposure of 1.8 mm fixed depth to guarantee adequate blood flow Electro-polished, silicone-coated needle with high puncture speed minimizes pain and facilitates healing for gentle finger tip sampling Easy-to-handle, retractable design features an intuitive, 2-step operation with low firing force and no lancet loading required