Learn More
Med Vet International McKesson Push Button Safety Lancet, 28 Gauge, 100/Box, 16-PBSL28GX, 20 Boxes
Catalog No. 503044333
Encompass_Preferred
Safety Lancet McKesson 28 Gauge Retractable Push Button Activation Finger Reliability provided by optimized needle exposure of 1.5 mm fixed depth to guarantee adequate blood flow Features gamma-sterilized 28-gauge needles, fully covered and protected with a sterility tab Electro-polished, silicone-coated needle with high puncture speed minimizes pain and facilitates healing for gentle Fingertiptip sampling Enhanced safety with sterility tab and self-destructing activation mechanism Purple color-coded design for easy gauge identification Disposable design for convenient single use
Catalog No. 50-304-4333 Supplier Med Vet International Supplier No. 1217991CS
Safety Lancet McKesson 28 Gauge Retractable Push Button Activation Finger Reliability provided by optimized needle exposure of 1.5 mm fixed depth to guarantee adequate blood flow Features gamma-sterilized 28-gauge needles, fully covered and protected with a sterility tab Electro-polished, silicone-coated needle with high puncture speed minimizes pain and facilitates healing for gentle Fingertiptip sampling Enhanced safety with sterility tab and self-destructing activation mechanism Purple color-coded design for easy gauge identification Disposable design for convenient single use