Learn More
Med Vet International Safety Lancet McKesson Prevent HP 21 Gauge Retractable Push Button Activation Finger, 16-PBHPSL21G1.8X, 100/Box
Catalog No. 503043153
Encompass_Preferred
Green in color, defines 21 needle gauge 1.8 mm depth Automatic needle retraction and lock out ensure sharp edge is enclosed before and after use Convenient, intuitive, one-step activation for precise and consistent results Color coded design for quick and easy identification Ready to use right out of the box; no set-up required Ergonomic, rounded shape and no-slip ribbing on grip make lancets easy to handle
Catalog No. 50-304-3153 Supplier Med Vet International Supplier No. 1217984BX
Green in color, defines 21 needle gauge 1.8 mm depth Automatic needle retraction and lock out ensure sharp edge is enclosed before and after use Convenient, intuitive, one-step activation for precise and consistent results Color coded design for quick and easy identification Ready to use right out of the box; no set-up required Ergonomic, rounded shape and no-slip ribbing on grip make lancets easy to handle