Learn More
Med Vet International McKesson Push Button Safety Lancet, 28 Gauge, 16-PBSL28GX, 100/Box
Catalog No. 503044305
Encompass_Preferred
Safety Lancet McKesson 28 Gauge Retractable Push Button Activation Finger Reliability provided by optimized needle exposure of 1.5 mm fixed depth to guarantee adequate blood flow Features gamma-sterilized 28-gauge needles, fully covered and protected with a sterility tab Electro-polished, silicone-coated needle with high puncture speed minimizes pain and facilitates healing for gentle Fingertiptip sampling Enhanced safety with sterility tab and self-destructing activation mechanism Purple color-coded design for easy gauge identification Disposable design for convenient single use
Catalog No. 50-304-4305 Supplier Med Vet International Supplier No. 1217991BX
Safety Lancet McKesson 28 Gauge Retractable Push Button Activation Finger Reliability provided by optimized needle exposure of 1.5 mm fixed depth to guarantee adequate blood flow Features gamma-sterilized 28-gauge needles, fully covered and protected with a sterility tab Electro-polished, silicone-coated needle with high puncture speed minimizes pain and facilitates healing for gentle Fingertiptip sampling Enhanced safety with sterility tab and self-destructing activation mechanism Purple color-coded design for easy gauge identification Disposable design for convenient single use