Learn More
Med Vet International Assure Safety Lancet, Sterile, 980128, 4800/Case
Catalog No. 503044223
Encompass_Preferred
100 / BX The hidden-needle construction and automatic after-use lockout helps prevent needlestick accidents Extended Finger Flange and Body Ridges: Engineered to provide a secure grip Single-turn Protective Cap: One, 360° turn and pull removes the protective cap Solid Body Color: Designed to make it easier to distinguish between different gauge sizes
Catalog No. 50-304-4223 Supplier Med Vet International Supplier No. 689752CS
100 / BX The hidden-needle construction and automatic after-use lockout helps prevent needlestick accidents Extended Finger Flange and Body Ridges: Engineered to provide a secure grip Single-turn Protective Cap: One, 360° turn and pull removes the protective cap Solid Body Color: Designed to make it easier to distinguish between different gauge sizes