Learn More
Med Vet International Lancet for Lancing Device McKesson 28 Gauge Non-Safety Twist Off Cap Finger, 16-028-100X, 100/Box, 50 Boxes
Catalog No. 503044775
Encompass_Preferred
Color: Blue 28 Gauge Needle Twist Top Sterile Tri-Bevel Tip Single use only. Universal design fits most lancing devices. Consistent depth penetration. Sterile, tri-beveled needle for added protection. Twist-activated safety device helps prevent needlestick injuries. Not made with natural rubber latex.
Catalog No. 50-304-4775 Supplier Med Vet International Supplier No. 1217978CS
Color: Blue 28 Gauge Needle Twist Top Sterile Tri-Bevel Tip Single use only. Universal design fits most lancing devices. Consistent depth penetration. Sterile, tri-beveled needle for added protection. Twist-activated safety device helps prevent needlestick injuries. Not made with natural rubber latex.