Learn More
Cayman Chemical Long-chain Fatty Acid Methyl Ester Mixture (porcine ovary), 100 mg
SDP
Catalog No. 502941060
Encompass_Preferred
A mixture of long-chain FAMEs; intended for use as a reference standard for the detection of these long-chain fatty acids in various sample types by GC-FID or GC-MS
Catalog No. 50-294-1060 Supplier Cayman Chemical Supplier No. 28662100 MG
A mixture of long-chain FAMEs; intended for use as a reference standard for the detection of these long-chain fatty acids in various sample types by GC-FID or GC-MS