Learn More
Cayman Chemical Long-chain Fatty Acid Methyl Ester Mixture (fish oil), 100 mg
SDP
Catalog No. 502938151
Encompass_Preferred
A mixture of long-chain FAMEs from fish oil; intended for use as a reference standard for the detection of these long-chain fatty acids in various sample types by GC-FID or GC-MS
Catalog No. 50-293-8151 Supplier Cayman Chemical Supplier No. 28661100 MG
A mixture of long-chain FAMEs from fish oil; intended for use as a reference standard for the detection of these long-chain fatty acids in various sample types by GC-FID or GC-MS