Learn More
Cayman Chemical LNgchaIn Faty AcId Methyl 50mg
SDP
Catalog No. 502937452
Encompass_Preferred
A mixture of long-chain methyl esters; contains five saturated and mono- and poly-unsaturated long-chain fatty acid methyl esters; intended for use as a reference standard for the detection of long-chain fatty acids in various sample types by GC-MS; supplied in an amber ampule in which the headspace has been purged with nitrogen/argon to prevent lipid oxidation
Catalog No. 50-293-7452 Supplier Cayman Chemical Supplier No. 2864550 MG
A mixture of long-chain methyl esters; contains five saturated and mono- and poly-unsaturated long-chain fatty acid methyl esters; intended for use as a reference standard for the detection of long-chain fatty acids in various sample types by GC-MS; supplied in an amber ampule in which the headspace has been purged with nitrogen/argon to prevent lipid oxidation