Learn More
Cayman Chemical LNgchaIn Saturated Faty Ac 1mL
SDP
Catalog No. 502988127
Encompass_Preferred
A mixture of five long-chain saturated fatty acid methyl esters; intended for use as a reference standard for the detection of long-chain saturated fatty acids in various samples by GC-MS; supplied in an amber ampule in which the headspace has been purged with nitrogen/argon to prevent lipid oxidation
Catalog No. 50-298-8127 Supplier Cayman Chemical Supplier No. 293571 ML
A mixture of five long-chain saturated fatty acid methyl esters; intended for use as a reference standard for the detection of long-chain saturated fatty acids in various samples by GC-MS; supplied in an amber ampule in which the headspace has been purged with nitrogen/argon to prevent lipid oxidation