Learn More
Cayman Chemical Long-chain Saturated Fatty Acid Methyl Ester Mixture 2, 1 mL
SDP
Catalog No. 502989081
Encompass_Preferred
A mixture of long-chain saturated FAMEs; intended for use as a reference standard for the quantification of these long-chain saturated fatty acids in various sample types by GC-FID or GC-MS
Catalog No. 50-298-9081 Supplier Cayman Chemical Supplier No. 293551 ML
A mixture of long-chain saturated FAMEs; intended for use as a reference standard for the quantification of these long-chain saturated fatty acids in various sample types by GC-FID or GC-MS