Learn More
Cayman Chemical Long-chain Saturated Fatty Acid Methyl Ester Mixture 1, 1 mL
SDP
Catalog No. 502978788
Encompass_Preferred
A mixture of long-chain saturated FAMEs; intended for use as a reference standard for the quantification of these long-chain saturated fatty acids in various sample types by GC-FID or GC-MS
Catalog No. 50-297-8788 Supplier Cayman Chemical Supplier No. 293521 ML
A mixture of long-chain saturated FAMEs; intended for use as a reference standard for the quantification of these long-chain saturated fatty acids in various sample types by GC-FID or GC-MS