Learn More
LONGER PUMP Pumps/ peristaltic pumps/ Longer Pump dPOFLEX PFU with pump head PFH02 Micro-volume Filling, Accuracy ±1%, CV <0.5%, filling volume 1uL-99.9L, Up to 32 Channels, FDA Compliant
Supplier:  LONGER PUMP 42/67
Encompass_Preferred
Pumps/ peristaltic pumps/ Longer Pump dPOFLEX PFUSpecifications- Pump head: PFH02- Filling volume: 1uL-99.99L- Filling accuracy: Better than ±1%- Filling precision: CV<0.5%- Filling time: 0.1 seconds-999.9 hours- Filling cycle: 0-999999, 0 means unlimited- Interval time: 0.1s - 999.9s- Back suction parameter: 0-30- Work mode: Single channel filling, multi-channel filling with the same parameters or individual parameters- Parameter recipe: Up to 500 recipes can be stored and recalled easily on the PFC controller- Communication control: RS485 interface, Modbus RTU protocol- External control input: Start the pump, emergency stop, disable channel- IP rating: IP31- Power supply: AC 100V-240V, 50/60Hz, 60W- Dimension: 119mm x 203mm x 176mm- Weight of system: 3.5kg- Working condition: Temperature: 5-40°C, RH: ≤85%, no condensation- Storage and transportation condition: Temperature: -40-70°C, RH: ≤85%, no condensation
Catalog No. NC2956862
Only null left
Add to Cart
Request a Quote