Learn More
Cayman Chemical Laninamivir octanoate, Purity: ≥98%, CAS: 203120-46-1, 50 mg
SDP
Catalog No. 502945014
Encompass_Preferred
An inhibitor of influenza neuraminidases and a prodrug form of laninamivir; selectively inhibits the neuraminidases from influenza A H1N1, H2N2, and H3N2 strains (mean IC50s = 822, 128, and 114 nM, respectively) over the neuraminidases from influenza B Victoria and Yamagata strains (mean IC50s = >10,000 nM for both); increases survival in a mouse model of lethal influenza H1N1 infection when administered intranasally at 0.95 µg/kg
Catalog No. 50-294-5014 Supplier Cayman Chemical Supplier No. 4085050 MG
An inhibitor of influenza neuraminidases and a prodrug form of laninamivir; selectively inhibits the neuraminidases from influenza A H1N1, H2N2, and H3N2 strains (mean IC50s = 822, 128, and 114 nM, respectively) over the neuraminidases from influenza B Victoria and Yamagata strains (mean IC50s = >10,000 nM for both); increases survival in a mouse model of lethal influenza H1N1 infection when administered intranasally at 0.95 µg/kg