Learn More
Cayman Chemical L-NMMA (acetate), Purity: ≥99%, CAS: 53308-83-1, 100 mg
SDP
Catalog No. 502890628
Encompass_Preferred
A non-selective NOS inhibitor (Kis = ~0.18, ~0.4, and ~6 µM for rat nNOS, human eNOS, and mouse iNOS, respectively)
Catalog No. 50-289-0628 Supplier Cayman Chemical Supplier No. 10005031100 MG
A non-selective NOS inhibitor (Kis = ~0.18, ~0.4, and ~6 µM for rat nNOS, human eNOS, and mouse iNOS, respectively)