Learn More
Cayman Chemical L-NMMA (acetate), Purity: ≥99%, CAS: 53308-83-1, 50 mg
SDP
Catalog No. 502890630
Encompass_Preferred
A non-selective NOS inhibitor (Kis = ~0.18, ~0.4, and ~6 µM for rat nNOS, human eNOS, and mouse iNOS, respectively)
Catalog No. 50-289-0630 Supplier Cayman Chemical Supplier No. 1000503150 MG
A non-selective NOS inhibitor (Kis = ~0.18, ~0.4, and ~6 µM for rat nNOS, human eNOS, and mouse iNOS, respectively)