Learn More
Cayman Chemical L-NMMA (acetate), Purity: ≥99%, CAS: 53308-83-1, 25 mg
SDP
Catalog No. 502888882
Encompass_Preferred
A non-selective NOS inhibitor (Kis = ~0.18, ~0.4, and ~6 µM for rat nNOS, human eNOS, and mouse iNOS, respectively)
Catalog No. 50-288-8882 Supplier Cayman Chemical Supplier No. 1000503125 MG
A non-selective NOS inhibitor (Kis = ~0.18, ~0.4, and ~6 µM for rat nNOS, human eNOS, and mouse iNOS, respectively)