Learn More
Cayman Chemical Linopirdine, Purity: ≥98%, CAS: 105431-72-9, 10 mg
SDP
Catalog No. 502982394
Encompass_Preferred
An enhancer of several neurotransmitters, including acetylcholine, dopamine, serotonin, and glutamate by blocking M-type K+ current (IC50 = 2.4 µM); also acts as an agonist of TRPV1 (EC50 = 115 µM in HEK293 cells voltage clamped at -60 mV)
Catalog No. 50-298-2394 Supplier Cayman Chemical Supplier No. 1954710 MG
An enhancer of several neurotransmitters, including acetylcholine, dopamine, serotonin, and glutamate by blocking M-type K+ current (IC50 = 2.4 µM); also acts as an agonist of TRPV1 (EC50 = 115 µM in HEK293 cells voltage clamped at -60 mV)