Learn More
Cayman Chemical LNP Dilution Buffer B (1X), 20 mL
SDP
Catalog No. 502942385
Encompass_Preferred
LNP dilution buffer B (1X) has been tested and formulated to work exclusively with Cayman’s LipidLaunch™ ALC-0315 LNP Kit (Loadable) (Item No. 702750).
Catalog No. 50-294-2385 Supplier Cayman Chemical Supplier No. 40077220 ML
LNP dilution buffer B (1X) has been tested and formulated to work exclusively with Cayman’s LipidLaunch™ ALC-0315 LNP Kit (Loadable) (Item No. 702750).