Learn More
Trajan Scientific LNR FOCUS TAPER HP S/S 5/PK
SDP
Catalog No. 502530186
Encompass_Preferred
SGE GC Inlet Tapered FocusLiners Split/Splitless, Diameter Inner: 0.4 cm, Diameter Outer: 0.63 cm, Length: 7.85 cm, For Use With: Agilent HP 5890, 6890, 7890, 6850, 4890, Thermo 9001, GCQ TRACE 1300, Varian 1177, Split/Splitless Tapered Focus Liner, Quantity: 5/Pk.
Catalog No. 50-253-0186 Supplier Trajan Scientific Supplier No. 092003
SGE GC Inlet Tapered FocusLiners Split/Splitless, Diameter Inner: 0.4 cm, Diameter Outer: 0.63 cm, Length: 7.85 cm, For Use With: Agilent HP 5890, 6890, 7890, 6850, 4890, Thermo 9001, GCQ TRACE 1300, Varian 1177, Split/Splitless Tapered Focus Liner, Quantity: 5/Pk.