Learn More
Trajan Scientific LNR FOCUS TAPER SHIM-17A 5/PK
SDP
Supplier:  Trajan Scientific 092068
Encompass_Preferred
Trajan GC Inlet Liners, Includes: Split/Splitless tapered FocusLiner, Diameter Inner: 3.4 mm, Diameter Outer: 5 mm, Overall Length: 95 mm, For Use With: 17 A and 2014 GC Inlet
Catalog No. 50-253-0178