Learn More
Trajan Scientific LNR HP 78.5X6.3X4MM TAPR 25/PK
SDP
Catalog No. 502530169
Encompass_Preferred
SGE Split/Splitless Liner with Single Taper, Diameter Inner: 0.4 cm, Diameter Outer: 0.63 cm, Length: 7.85 cm, For Use With: Agilent HP 5890, 6890, 7890, 6850, 4890, Thermo 9001, GCQ TRACE 1300, Varian 1177, Includes: Single taper, GC Split/Splitless Liner, Quantity: 25/Pk.
Catalog No. 50-253-0169 Supplier Trajan Scientific Supplier No. 092229
SGE Split/Splitless Liner with Single Taper, Diameter Inner: 0.4 cm, Diameter Outer: 0.63 cm, Length: 7.85 cm, For Use With: Agilent HP 5890, 6890, 7890, 6850, 4890, Thermo 9001, GCQ TRACE 1300, Varian 1177, Includes: Single taper, GC Split/Splitless Liner, Quantity: 25/Pk.