Learn More
Trajan Scientific LNR HP 78.5X6.3X4MM TPR 5PK
SDP
Catalog No. 502550759
Encompass_Preferred
SGE Splitless Liner with Single Taper and Quartz Wool, Diameter Inner: 0.4 cm, Diameter Outer: 0.63 cm, Length: 7.85 cm, For Use With: Agilent HP 5890, 6890, 7890, 6850, 4890, Thermo 9001, GCQ TRACE 1300, Varian 1177, Includes: Single taper, Packing Material: Quartz Wool, GC Splitless Liner
Catalog No. 50-255-0759 Supplier Trajan Scientific Supplier No. 092019
SGE Splitless Liner with Single Taper and Quartz Wool, Diameter Inner: 0.4 cm, Diameter Outer: 0.63 cm, Length: 7.85 cm, For Use With: Agilent HP 5890, 6890, 7890, 6850, 4890, Thermo 9001, GCQ TRACE 1300, Varian 1177, Includes: Single taper, Packing Material: Quartz Wool, GC Splitless Liner