Learn More
Trajan Scientific LNR HPFCSLNR 78.5X6.3X4MM 5/PK
SDP
Catalog No. 502550760
Encompass_Preferred
SGE GC Inlet FocusLiners, Quantity: 5/Pk., Diameter Inner: 0.4 cm, Diameter Outer: 0.63 cm, Length: 7.85 cm, For Use With: Agilent HP 5890, 6890, 7890, 6850, 4890, Thermo 9001, GCQ TRACE 1300, Varian 1177, Packing Material: Quartz Wool, Split/Splitless Fast Focus Liner
Catalog No. 50-255-0760 Supplier Trajan Scientific Supplier No. 092002
SGE GC Inlet FocusLiners, Quantity: 5/Pk., Diameter Inner: 0.4 cm, Diameter Outer: 0.63 cm, Length: 7.85 cm, For Use With: Agilent HP 5890, 6890, 7890, 6850, 4890, Thermo 9001, GCQ TRACE 1300, Varian 1177, Packing Material: Quartz Wool, Split/Splitless Fast Focus Liner