Learn More
Trajan Scientific LNR INL FCS AGLNT HP2.3MM 5/PK
SDP
Catalog No. 502530174
Encompass_Preferred
SGE Split/Splitless Tapered Fast FocusLiner, Diameter Inner: 0.23 cm, Diameter Outer: 0.63 cm, Length: 7.85 cm, For Use With: Agilent HP 5890, 6890, 7890, 6850, 4890, Thermo 9001, GCQ TRACE 1300, Varian 1177, Packing Material: Quartz Wool, Split/Splitless Tapered Fast Focus Liner, Quantity: 5/Pk.
Catalog No. 50-253-0174 Supplier Trajan Scientific Supplier No. 092111
SGE Split/Splitless Tapered Fast FocusLiner, Diameter Inner: 0.23 cm, Diameter Outer: 0.63 cm, Length: 7.85 cm, For Use With: Agilent HP 5890, 6890, 7890, 6850, 4890, Thermo 9001, GCQ TRACE 1300, Varian 1177, Packing Material: Quartz Wool, Split/Splitless Tapered Fast Focus Liner, Quantity: 5/Pk.