Learn More
Trajan Scientific LNR INL TQCE 2MMD TRC PTV 5/PK
SDP
Catalog No. 502530173
Encompass_Preferred
Trajan Thermo Scientific Trace and Focus GC Liners, Diameter Inner: 1.75 mm, Diameter Outer: 2.75 mm, Length: 120 mm, For Use With: Thermo Scientific Trace and Focus GCs, GC Inlet Liner, PTV Liner, Quantity: 5/Pk.
Catalog No. 50-253-0173 Supplier Trajan Scientific Supplier No. 092142
Trajan Thermo Scientific Trace and Focus GC Liners, Diameter Inner: 1.75 mm, Diameter Outer: 2.75 mm, Length: 120 mm, For Use With: Thermo Scientific Trace and Focus GCs, GC Inlet Liner, PTV Liner, Quantity: 5/Pk.