Learn More
Trajan Scientific LNR PE AUTOSYS FCSLNR 3MM 5/PK
SDP
Catalog No. 502530177
Encompass_Preferred
Trajan GC Inlet Liners, Includes: Split/Splitless FocusLiner; For AutoSystem and Clarus 500, Diameter Inner: 4 mm, Diameter Outer: 6.2 mm, Overall Length: 92 mm, Quantity: 5/Pk.
Catalog No. 50-253-0177 Supplier Trajan Scientific Supplier No. 092092
Trajan GC Inlet Liners, Includes: Split/Splitless FocusLiner; For AutoSystem and Clarus 500, Diameter Inner: 4 mm, Diameter Outer: 6.2 mm, Overall Length: 92 mm, Quantity: 5/Pk.