Learn More
Trajan Scientific LNR SHIMADZU17A 3MMFCSLNR 5/PK
SDP
Catalog No. 502530181
Encompass_Preferred
Trajan GC Inlet Liners, Includes: Split/Splitless FocusLiner, Diameter Inner: 3.4 mm, Diameter Outer: 5 mm, Overall Length: 95 mm, For Use With: 17 A and 2014 GC Inlet
Catalog No. 50-253-0181 Supplier Trajan Scientific Supplier No. 092062
Trajan GC Inlet Liners, Includes: Split/Splitless FocusLiner, Diameter Inner: 3.4 mm, Diameter Outer: 5 mm, Overall Length: 95 mm, For Use With: 17 A and 2014 GC Inlet