Learn More
Trajan Scientific LNR VA SPI 54X4.7X0.8MM 5/PK
SDP
Catalog No. 502530182
Encompass_Preferred
Liners, Varian/Bruker; SGE; SPI liner; Restriction: 0.5mm; For: 0.53mm ID On-column; OD: 4.6mm; ID: 0.8mm; Length: 54mm; Pk size: 5
Catalog No. 50-253-0182 Supplier Trajan Scientific Supplier No. 092034
Liners, Varian/Bruker; SGE; SPI liner; Restriction: 0.5mm; For: 0.53mm ID On-column; OD: 4.6mm; ID: 0.8mm; Length: 54mm; Pk size: 5