Learn More
Cayman Chemical Lansoprazole-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 934294-22-1, 1 mg
SDP
Catalog No. 502932535
Encompass_Preferred
An internal standard for the quantification of lansoprazole by GC- or LC-MS
Catalog No. 50-293-2535 Supplier Cayman Chemical Supplier No. 252271 MG
An internal standard for the quantification of lansoprazole by GC- or LC-MS