Learn More
Cayman Chemical Lnsoprazole sulfon 10mg
SDP
Catalog No. 502986896
Encompass_Preferred
A metabolite of lansoprazole; formed by metabolism of lansoprazole by CYP3A4
Catalog No. 50-298-6896 Supplier Cayman Chemical Supplier No. 2256910 MG
A metabolite of lansoprazole; formed by metabolism of lansoprazole by CYP3A4