Learn More
Bellco Glass, Inc. Lo/Profile Dual Pan Hot Shaker, High Speed, 115V
SDP
Supplier:  Bellco Glass, Inc. 774652115
Encompass_Preferred
Bellco’s new Dual Pan Low Profile Hot Shaker provides gentle, large orbit shaking motion combined with precise temperature control. Designed to be the perfect solution for incubation of electrophoretic gels and filter hybridization media, the Low Profile layout promotes easy access to the independent microprocessor driven temperature controller and reliable control of shaking speeds up to 75 oscillations per minute. The quiet brushless drive motor and repeatable, stable temperature settings from 5°C above ambient up to 99.9°C may be effectively used in most biochemical, chemical and microbiological applications.. MADE IN THE USA
Catalog No. 50-194-6180
Add to Cart
Request a Quote